Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Disodium L-(+)-Tartrate DihydrateL(+)-Sodium tartrate dibasic dihydrateSodium L-tartrate dibasic dihydrate(R,R)-(+)-Sodium tartrate dibasic dihydrateL(+)-Tartaric acid disodium salt dihydrate(R,R)-(+)-Disodium tartrate dihydrateSodium Tartrate, DihydrateL(+)-Tartaric acid, disodium salt, dihydrate(R,R)-(+)-Tartaric acid disodium salt dihydrate(+)-Disodium L-tartrate dihydrate(+)-Disodium-L-tartrat dihydrateSodium L-(+)-tartrate dihydrate

Management of Change (MOC) category = N

MACR2386-20 MACR2386-04 MACR2386-20
MK238620EA 4060.82 USD InStock
MK238620 MK238604 CAMK238620
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
di-Sodium L(+)-tartrate dihydrate


Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57 °C
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/17/2299817.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂C₄H₄O₆·2H₂O) 99.0 - 101.0 %
ACS - Loss on Drying at 150°C 15.61 - 15.71 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤5 ppm
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonium (NH₄) ≤0.003 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty