Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 30%, Technical Grade for trace metal analysis (20° Baume), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Muriatic acid

Management of Change (MOC) category = N

MACR2624-44 MACR2624-44
MK262444CS 406 USD InStock
MK262444 CAMK262444
Hydrochloric acid 30%, Technical Grade for trace metal analysis (20° Baume), Macron Fine Chemicals™
Hydrochloric acid
Technical - Chemicals of selected commercial quality, useful in some applications.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (HCl) ≥34 %
Identification Passes Test
Arsenic (As) ≤5 ppm
Free Chlorine (as Cl) ≤1 ppm
Nitrate (NO₃) ≤0.005 %
Other Heavy Metals (as Pb) ≤1 ppm
Total Iron (Fe) ≤10 ppm
Appearance (clear, colorless to pale yellow or green solution)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty