Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 69.0 - 72.0%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

MACR2766-14 MACR2766-14 2766-44 MACR2766-44
MK276614CS 1560.41 USD InStock
MK276614 MK2766-44 CAMK276614 CAMK2766-44
Perchloric acid 69.0 - 72.0%, AR® ACS, Macron Fine Chemicals™
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112.2 °C
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HClO₄) 69.0 - 72.0 %
Appearance (clear, colorless solution) Passes Test
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Color (APHA) ≤ 10
ACS - Heavy Metals (by ICP-OES) ≤ 1 ppm
ACS - Iron (Fe) ≤ 0.0001 %
ACS - Nitrogen Compounds (as N) ≤ 0.001 %
ACS - Residue after Ignition ≤ 0.003 %
ACS - Silicate and Phosphate (as SiO₂) ≤ 0.0005 %
ACS - Sulfate (SO₄) ≤ 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty