Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran ≥99.8% contains no preservatives, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Butylene oxideOxolaneTHFCyclotetramethylene oxideTetramethylene oxideDiethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

Management of Change (MOC) category = N

MACR2858-08 MACR2858-06 MACR2858-06
MK285808CS 1143.63 USD InStock
MK285808 MK285806 CAMK285806
Tetrahydrofuran ≥99.8% contains no preservatives, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Tetrahydrofuran
ChromAR—Solvents that meet ACS specifications and are suitable for liquid chromatography and UV-spectrophotometry. 


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99.8 %
Peroxide (as H₂O₂) ≤0.015 %
Residue after Evaporation ≤5 ppm
Water (H₂O)(by KF, coulometric) ≤0.03 %
Optical Absorbance (1-cm path vs water) - 212 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.17
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.01
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty