Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium sulfate, Drierite™ (8 Mesh), with indicator (cobalt (II) chloride), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

MACR3378-04
MK337804CS 2138.96 USD InStock
MK337804
Calcium sulfate, Drierite™ (8 Mesh), with indicator (cobalt (II) chloride), Macron Fine Chemicals™
Calcium sulphate


Formula: CaO₄S
MW: 136.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty