Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: PhenylamineAminobenzene

Management of Change (MOC) category = N

MACR3584-08 3584-04 MACR3584-08 MACR3584-04
MK358404CS 2293.61 USD InStock
CAMK358408 MK358404 CAMK358404 MK358408
Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
Aniline


Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: –6.2 °C
Density: 1.019 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/50/758750.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅NH₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) (at time of packaging) ≤250
Residue after Ignition ≤0.005 %
Chlorobenzene (C₆H₅Cl) ≤0.01 %
Hydrocarbons Passes Test
Nitrobenzene (C₆H₅NO₂) Passes Test
NOTE: This reagent darkens to a reddish-brown color on storage.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty