Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Slaked lime

MACR4188-40 MACR4188-25 MACR4188-24 MACR4188-05 MACR4188-05 MACR4188-25 MACR4188-24 MACR4188-05 MACR4188-24 MACR4188-40
JT4188-05CS 217.25 USD InStock
JT4188-05 MK418840 CAMK418825 JT4188-24 CAMK4188-05 MK418825 CAMK4188-24 MK4188-05 CAMK418840 MK4188-24
Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™
Calcium hydroxide


Formula: Ca(OH)₂
MW: 74.09 g/mol
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (Ca(OH)₂) (by EDTA titrn) 95.0 - 100.5 %
Appearance Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Carbonate (CO₃) Passes Test
FCC - Acid-Insoluble Substances ≤0.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤2 ppm
FCC - Fluoride (F) ≤0.005 %
FCC - Magnesium and Alkali Salts ≤4.8 %
Thru U.S. No. 325 Sieve, For Information Only
Barium (Ba), For Information Only

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty