Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium oxide 95.0-100.5% FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Lime

MACR4268-24 MACR4268-24
MK4268-24EA 544.78 USD InStock
CAMK4268-24 MK4268-24
Calcium oxide 95.0-100.5% FCC, Macron Fine Chemicals™
Calcium oxide


Formula: CaO
MW: 56.08 g/mol
MDL Number: MFCD00010911
CAS Number: 1305-78-8
UN: 1910
ADR: 8,III
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (CaO) 95.0 - 100.5 %
FCC - Identification Passes Test
FCC - Acid-Insoluble Substances ≤1 %
FCC - Alkalies or Magnesium ≤3.6 %
FCC - Arsenic (As) ≤3 mg/kg
FCC - Fluoride (F) ≤0.015 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Ignition ≤10.0 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty