Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium phosphate tribasic, powder NF, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: tri-Calcium phosphateCalcium Hydroxide PhosphateHApHydroxylapatiteCalcium hydroxyphosphateTribasic calcium phosphate

Management of Change (MOC) category = R

MACR4280-24 MACR4280-03 MACR4280-03 MACR4280-24
MK428024EA 1905.47 USD InStock
MK428024 MK428003 CAMK428003 CAMK428024
Calcium phosphate tribasic, powder NF, FCC, Macron Fine Chemicals™
Calcium phosphate tribasic


Formula: Ca₅HO₁₃P₃
MW: 502.306 g/mol
Density: 3.14 g/cm³
MDL Number: MFCD00010904
CAS Number: 1306-06-5
Order Now
Specification Test Results
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Assay (Ca) 34.0 - 40.0 %
NF - Acid-Insoluble Substances ≤ 0.2 %
NF - Arsenic (As) ≤ 3 mg/kg
NF - Barium (Ba) Passes Test
NF - Carbonate (CO₃) Passes Test
NF - Chloride (Cl) ≤ 0.14 %
NF - Dibasic Salt and Calcium Oxide (ml) 13.0 - 14.3
NF - Fluoride (F) ≤ 0.0075 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Loss on Ignition ≤ 8.0 %
NF - Nitrate (NO₃) Passes Test
NF - Sulfate (SO₄) ≤ 0.8 %
NF - Water-Soluble Substances ≤ 0.5 %
FCC - Assay (Ca) 34.0 - 40.0 %
FCC - Arsenic (As) ≤ 3 mg/kg
FCC - Fluoride (F) ≤ 0.0075 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Loss on Ignition ≤ 8.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty