Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium sulfate 97.5-100.5% USP suitable for use as excipient, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = R

MACR4518-26 MACR4518-26
MK451826EA 3906.71 USD InStock
MK451826 CAMK451826
Barium sulfate 97.5-100.5% USP suitable for use as excipient, Macron Fine Chemicals™
Barium sulphate


Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1345 °C
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (BaSO₄) 97.5 - 100.5 %
USP - Acid-Soluble Substances ≤0.3 %
USP - Bulk (fl. oz/lb) 20 - 31
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (10% Suspension) 3.5 - 10.0
USP - Soluble Barium Salts ≤0.001 %
USP - Sulfide (S) ≤0.5 ppm
Suspension Test for Dirt Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty