Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate 0.1 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

Management of Change (MOC) category = NI

MACR4682-01 MACR4682-60 MACR4682-01
MK468201CS 462.44 USD InStock
MK468201 MK468260 CAMK468201
Sodium thiosulfate 0.1 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Sodium thiosulphate


Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty