Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) chloride dihydrate ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Cupric chloride dihydrateCopper dichloride dihydrate

Management of Change (MOC) category = N

MACR4824-20 MACR4824-20
MK482420EA 8067.31 USD InStock
MK482420 CAMK482420
Copper(II) chloride dihydrate ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™
Copper (II) chloride dihydrate


Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CuCl₂·2H₂O) (by iodometry) ≥99.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.005 %
ACS - Nitrate (NO₃) ≤0.015 %
Solubility Passes Test
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Sodium (Na) ≤0.02 %
ACS - Nickel (Ni) ≤0.01 %
ACS - Potassium (K) ≤0.01 %
ACS - Calcium (Ca) ≤0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty