Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexane ≥99.0% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Hexahydro-BenzeneHexahydrobenzeneHexamethylene

Management of Change (MOC) category = N

MACR4878-22 MACR4878-22 4878-16 MACR4878-02 MACR4878-16 MACR4878-02
MK487816CS 1446.01 USD InStock
CAMK487822 MK487816 CAMK487802 CAMK487816 MK487822 MK487802
Cyclohexane ≥99.0% (by GC), AR® ACS, Macron Fine Chemicals™
Cyclohexane


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₁₂) (by GC, corrected for water) ≥99.0 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Water (H₂O)(by KF, coulometric) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty