Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methane dichlorideMethylene chloride

Management of Change (MOC) category = N

MACR4879-06 MACR4879-10 MACR4879-10
MK487906CS 520.34 USD InStock
CAMK487910 MK487906 MK487910
Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™
Dichloromethane
Suitable for liquid chromatography and UV-spectrophotometry.


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.9 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0003 %
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Free Halogens Passes Test
ACS - Water (H₂O)(by coulometry) ≤0.01 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm ≤1.0
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.12
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 - 400 nm ≤0.01
Liquid Chromatography Suitability Absorbance Passes Test
Appearance (clear, colorless liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty