Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride, anhydrous 93.0-100.5%, CALTAC FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Chloro calcium

MACR4880-25 MACR4880-40 MACR4880-32 MACR4880-40 MACR4880-32 MACR4880-25 MACR4880-55
MK4880-40EA 4614.45 USD InStock
MK4880-40 MK488025 CAMK4880-55 CAMK488025 CAMK4880-40 MK4880-32 JT4880-32
Calcium chloride, anhydrous 93.0-100.5%, CALTAC FCC, Macron Fine Chemicals™
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 13,01660
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (CaCl₂) 93.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
Solubility Passes Test
Acid-Insoluble Substances - Retained Mass ≤0.02 %
Acid-Insoluble Substances - No Particles >2mm Passes Test
Arsenic (As) ≤3 mg/kg
Fluoride (F) ≤0.004 %
FCC - Lead (Pb) ≤5 mg/kg
Magnesium and Alkali Salts ≤5.0 %
pH (1 in 20) 4.5 - 11.0

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty