Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Methane dichlorideMethylene chloride

Management of Change (MOC) category = N

MACR4881-04 MACR4881-06 MACR4881-26 MACR4881-08 MACR4881-19 MACR4881-04 MACR4881-06 MACR4881-08 MACR4881-26 MACR4881-19
MK488104CS 1098.28 USD InStock
CAMK488119 CAMK488108 MK488104 MK488126 MK488106 MK488108 MK488119 CAMK488104 CAMK488126 CAMK488106
Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC) ≥99.5 %
Appearance (clear, colorless liquid) Passes Test
ACS - Carbon Tetrachloride (mg/L) ≤10.0000 ppm
ACS - Color (APHA) ≤10
ACS - Free Halogens Passes Test
ACS - Residue after Evaporation ≤0.002 %
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Water (H₂O)(by coulometry) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty