Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Histidine 98.5-101.5% (dried basis) USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: L-3-Imidazol-4-ylalanine(S)-(+)-Histidine(S)-2-Amino-3-(4-imidazolyl)propionic acidNSC 137773L-HistidineH-His-OH

Management of Change (MOC) category = R

MACR4928-07 MACR4928-06 MACR4928-06 MACR4928-07
MK4928-06EA 2823.22 USD InStock
MK4928-06 MK4928-07 CAMK4928-07 CAMK4928-06
L(+)-Histidine 98.5-101.5% (dried basis) USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L(+)-Histidine


Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD00064315
CAS Number: 71-00-1
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Store in airtight container.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₉N₃O₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ D (+) 12.6 - 14.0 Degree
USP - pH 7.0 - 8.5
USP - Loss on Drying at 105°C ≤ 0.2 %
USP - Residue on Ignition ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₆H₉N₃O₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²ºD (+) 11.4 - 12.4 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C6H9N3O2) (dried basis) 99.0 - 101.0 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Chloride (Cl) ≤ 0.021 %
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Iron (Fe) ≤ 10 ppm
JP - Identification Passes Test
JP - Loss on Drying at 105°C ≤ 0.30 %
JP- Optical Rotation [Å]²ºD (+) 11.8 - 12.8 Degree
JP - Related Substances Passes Test
JP - pH 7.0 - 8.5
JP - Residue on Ignition ≤ 0.10 %
JP - Sulfate (SO₄) ≤ 0.028 %
ChP - Assay (dried basis) 99.0 - 101.0 %
ChP -Specific Rotation @ 20 °C (+) 12.0 - 12.8 Degree
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Acidity or alkalinity (pH) 7.0 - 8.5
ChP - Transmittance of solution ≥ 98.0 %
ChP - Chlorides ≤ 0.02 %
ChP - Sulfates ≤ 0.02 %
ChP - Ammonium ≤ 0.02 %
ChP - Other amino acids (TLC)(<lt/≥0.5%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤ 0.2 %
ChP - Residue on ignition ≤ 0.1 %
ChP - Iron ≤ 0.001 %
ChP - Heavy metals ≤ 0.001 %
ChP - Arsenic ≤ 0.0001 %
ChP - Bacterial endotoxin, EU/g ≤ 6.0
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty