Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Arginine 98.5-101.5% (dried basis) USP, FCC, Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: L-ArginineH-L-Arg-OHArginine(S)-(+)-ArginineH-Arg-OH(S)-2-Amino-5-guanidinopentanoic acid

Management of Change (MOC) category = R

MACR4932-09 MACR4932-07 MACR4932-06 MACR4932-06 MACR4932-07 MACR4932-05 MACR4932-09 MACR4932-05
MK4932-06EA 865.82 USD InStock
CAMK493207 MK4932-06 CAMK4932-09 MK4932-05 MK493207 CAMK4932-05 MK4932-09 CAMK4932-06
L(+)-Arginine 98.5-101.5% (dried basis) USP, FCC, Multi-Compendial, Macron Fine Chemicals™
L(+)-Arginine


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
MDL Number: MFCD00002635
CAS Number: 74-79-3
Order Now
Specification Test Results
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₁₄N₄O₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 26.3 - 27.7 Degree
USP - Loss on Drying at 105°C ≤ 0.5 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Residue on Ignition ≤ 0.3 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₆H₁₄N₄O₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 25.5 - 28.5 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Identification Passes Test
JP - Optical rotation, 20° C (+) 26.9 - 27.9 Degree
JP - pH (1:10) 10.5 - 12.0
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Iron (Fe) ≤ 10 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤ 0.30 %
JP - Residue after Ignition ≤ 0.1 %
JP - Assay (C₆H₁₄N₄O₂)(dried basis) 98.5 - 101.0 %
Endotoxin Concentration, IU/mg, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty