Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Valine 98.5-101.5% (dried basis) USP, FCC, Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (S)-(+)-ValineValineH-Val-OH

MACR4936-09 MACR4936-09 MACR4936-06 MACR4936-06
MK4936-06EA 2152.99 USD InStock
MK4936-06 CAMK4936-06 CAMK4936-09 MK4936-09
L(+)-Valine 98.5-101.5% (dried basis) USP, FCC, Multi-Compendial, Macron Fine Chemicals™
L(+)-Valine


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Density: 1.23 g/cm³
MDL Number: MFCD00064220
CAS Number: 72-18-4
Order Now
Specification Test Results
Meets B.P. Chemical Specifications
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 26.6 - 28.8 Degree
USP - pH 5.5 - 7.0
USP - Loss on Drying at 105°C ≤0.3 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.05 %
USP - Sulfate (SO₄) ≤0.03 %
USP - Iron (Fe) ≤0.003 %
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
EP/BP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 26.5 - 29.0 Degree
EP/BP-Ninhydrin-Positive Substances-Impurity B ≤0.4 %
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤1.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤0.02 %
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (C₅H₁₁NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 26.5 - 29.0 Degree
JP - pH 5.5 - 6.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021 %
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.30 %
JP - Residue on Ignition ≤0.10 %
Endotoxin Concentration, IU/mg, For Information Only
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty