Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Histidine monohydrochloride monohydrate 98.5-101.5% (dried basis), Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: H-His-OH.HCl.H2OL-Histidine monohydrochloride monohydrate(S)-(+)-Histidine monohydrochloride monohydrateL-Histidine hydrochloride hydrateHis HCl monohydrateH-His-OH·HCl·H₂O(2S)-(+)-Histidine monohydrochloride monohydrateL-Histidine hydrochloride monohydrateL-3-Imidazol-4-ylalanine monohydrochloride monohydrate

Management of Change (MOC) category = R

MACR4942-05 MACR4942-07 MACR4942-05 MACR4942-07 MACR4942-06 MACR4942-06 MACR4942-09 MACR4942-09
MK4942-05EA 151.79 USD InStock
MK4942-05 CAMK494207 CAMK4942-05 MK4942-06 MK4942-09 MK494207 CAMK4942-09 CAMK4942-06
L(+)-Histidine monohydrochloride monohydrate 98.5-101.5% (dried basis), Multi-Compendial, Macron Fine Chemicals™
L(+)-Histidine monohydrochloride monohydrate


Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
MDL Number: MFCD00151027
CAS Number: 5934-29-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/30/2299830.jpg
Order Now
Specification Test Results
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
EP/BP - Assay (C₆H₉N₃O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification F Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 3.0 - 5.0
EP/BP - Specific Rotation [Å]²º^D (+) 9.2 - 10.6 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying at 150°C 7.0 - 10.0 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Identification Passes Test
JP - Optical Rotation (+) 9.2 - 10.6
JP - pH 3.5 - 4.5
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Iron (Fe) ≤ 10 ppm
JP - Related Substances Passes Test
JP - Water 7.2 - 10.0 %
JP - Residue on Ignition ≤ 0.10 %
JP - Assay (anhydrous basis) 99.0 - 101.0 %
Endotoxin Concentration, IU/mg ,For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty