Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Isoleucine USP, Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (2S,3S)-(+)-IsoleucineH-Ile-OHL-Isoleucine

MACR4943-07 MACR4943-07 MACR4943-05 MACR4943-06 MACR4943-05 MACR4943-06
MK4943-07EA 28830.84 USD InStock
MK4943-07 CAMK4943-05 CAMK4943-06 MK4943-05 MK4943-06 CAMK4943-07
L(+)-Isoleucine USP, Multi-Compendial, Macron Fine Chemicals™
L(+)-Isoleucine
L-Isoleucine, U.S.P. Multi-Compendial. GMP Manufactured Product Bulk Pharmaceutical Chemical

Only Class 2 Solvents (Methanol) are likely to be present. All are below Option 1 limits.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impuri profile report.

Caution: For Manufacturing, processing or repackaging


Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 13,05198
Order Now
Specification Test Results
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
GMP Manufactured Product
USP - Assay (C₆H₁₃NO₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 38.9 - 41.8 °
USP - pH 5.5 - 7.0
USP - Loss on Drying at 105°C ≤ 0.3 %
USP - Residue on Ignition ≤ 0.3 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₆H₁₃NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 40.0 - 43.0 °
EP/BP-Ninhydrin-Positive Substances-Impurity A ≤ 0.3 %
EP/BP-Ninhydrin-Positive Substances-Impurity C ≤ 0.3 %
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 1.0 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₆H₁₃NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) 39.5 - 41.5 °
JP - pH 5.5 - 6.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤ 0.30 %
JP - Residue on Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg, For Information Only < 0.01

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty