Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Phenylalanine 98.5-101.5% (dried basis) USP, Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (S)-(-)-PhenylalanineH-L-Phe-OHPhenylalanineH-Phe-OHL-α-Phenylalanine3-Phenyl-L-alanine3-Phenyl-L(-)-alanine

Management of Change (MOC) category = R

MACR4945-06 MACR4945-07 MACR4945-07 MACR4945-06
MK4945-06EA 1822.69 USD InStock
MK4945-06 CAMK4945-07 CAMK4945-06 MK4945-07
L-Phenylalanine 98.5-101.5% (dried basis) USP, Multi-Compendial, Macron Fine Chemicals™
L-α-Phenylalanine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Order Now
Specification Test Results
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₉H₁₁NO₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) -34.7 - -32.7 Degree
USP - pH 5.4 - 6.0
USP - Loss on Drying at 105°C ≤ 0.3 %
USP - Residue on Ignition ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₉H₁₁NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) -35.5 - -33.0 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₉H₁₁NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation (+) -35.5 - -33.0 Degree
JP - pH 5.3 - 6.3
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤ 0.30 %
JP - Residue on Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg, For Information Only
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty