Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Methyl sulfoxideDMSOMethyl sulphoxide

Management of Change (MOC) category = N

MACR4948-16 MACR4948-16 MACR4948-02
MK494816CS 3073.62 USD InStock
CAMK494816 MK494816 MK494802
Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃)₂SO) (by GC, corrected for water) ≥99.9 %
Appearance (clear, colorless liquid) Passes Test
ACS - Residue after Evaporation ≤0.01 %
ACS - Titrable Acid (meq/g) ≤0.001
ACS - Water (H₂O)(by coulometry) ≤0.1 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty