Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

Management of Change (MOC) category = N

MACR5092-02 MACR5092-16 MACR5092-02 MACR5092-16
MK509202CS 1784.14 USD InStock
CAMK509216 CAMK509202 MK509202 MK509216
Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Glycerine


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₃H₅(OH)₃) (by GC, corrected for water) ≥99.5 %
ACS - Acrolein and Glucose Passes Test
ACS - Chlorinated Compounds (as Cl) ≤0.003 %
ACS - Color (APHA) ≤10
ACS - Fatty Acid Esters (as Butyric Acid) ≤0.05 %
ACS - Heavy Metals (as Pb) ≤2 ppm
ACS - Neutrality Passes Test
ACS - Residue after Ignition ≤0.005 %
Solubility (5 in 50) Passes Test
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Appearance (clear, colorless, syrupy liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty