Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium sulfate heptahydrate 99.0-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: sulfuric acid magnesium salt heptahydrateBitter saltsulphuric acid magnesium salt heptahydrateEpsom salt

Management of Change (MOC) category = R

MACR5691-25 MACR5691-24 MACR5691-24 MACR5691-25
MK569124EA 3107.61 USD InStock
CAMK5691-25 MK569124 CAMK569124 MK5691-25
Magnesium sulfate heptahydrate 99.0-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™
Magnesium sulphate heptahydrate


Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715
Order Now
Specification Test Results
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (as MgSO₄) 99.0 - 100.5 %
Appearance Passes Test
Solubility (5 in 50) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH 5.0 - 9.2
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Selenium (Se) ≤ 0.003 %
USP - Iron (Fe) ≤ 0.5 ppm
Assay (MgSO₄) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Chloride (Cl) ≤ 300 ppm
EP - Iron (Fe) ≤ 20 ppm
EP/BP - Loss on Drying 48.0 - 52.0 %
Elemental Impurities (USP (232), EP 5.20) - Information on
elemental impurities for this product is available on the
associated Product Regulatory Data Sheet and elemental impurity
profile report.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty