Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = N

MACR5840-06 MACR5840-02 MACR5840-02 MACR5840-06
MK584002CS 2028.05 USD InStock
CAMK5840-06 MK584002 CAMK584002 MK5840-06
Lithium carbonate ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Lithium carbonate


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Li₂CO₃) ≥99.0 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals ≤0.002 %
ACS - Insoluble in Dilute HCl ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Nitrate (NO₃) ≤5 ppm
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.1 %
ACS - Sulfur Compounds (as SO₄) ≤0.2 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty