Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium carbonate 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Magnesium Carbonate

Management of Change (MOC) category = R

MACR5942-32 MACR5942-32
MK594232EA 748.58 USD InStock
MK594232 CAMK594232
Magnesium carbonate 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™
Magnesium carbonate


Formula: CMgO₃
MW: 84.31 g/mol
Density: 3.03 g/cm³ (16 °C)
MDL Number: MFCD00064632
CAS Number: 546-93-0
EINECS: 208-915-9
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (MgO) 40.0 - 43.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Microbiological Passes Test
USP - Soluble Salts ≤ 1.0 %
USP - Acid-Insoluble Substances ≤ 0.05 %
USP - Arsenic (As) ≤ 3 ppm
USP - Limit of Calcium (Ca) ≤ 0.45 %
USP - Iron (Fe) ≤ 0.02 %
FCC - Assay (MgO) 40.0 - 43.5 %
FCC - Identification Passes Test
FCC - Soluble Salts ≤ 1.0 %
FCC - Acid-Insoluble Substances ≤ 0.05 %
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Calcium Oxide ≤ 0.6 %
Appearance (bulky, white powder) Passes Test
Solubility in Dilute Sulfuric Acid Passes Test
Residue on US 325 Mesh ≤ 0.3 %
Tapped Density ≥ 0.43 g/ml
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty