Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium hydroxide 95.0-100.5%, white powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = R

MACR5984-20 MACR5984-12 MACR5984-12 MACR5984-20 MACR5984-25 MACR5984-25
MK598420EA 3208.67 USD InStock
MK598420 CAMK5984-12 MK5984-12 MK5984-25 CAMK598420 CAMK5984-25
Magnesium hydroxide 95.0-100.5%, white powder USP, FCC, Macron Fine Chemicals™
Magnesium hydroxide


Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.3261…2.3631 g/cm³ (20 °C)
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (Mg(OH)₂) (dried basis) 98.0 - 102.0 %
USP - Calcium (Ca) ≤ 1.5 %
USP - Carbonate (CO₃) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
Lead (Pb)(USP) ≤ 1.5 ppm
USP - Loss on Drying at 105°C ≤ 2.0 %
USP - Loss on Ignition 30.0 - 33.0 %
USP - Soluble Salts ≤ 2.0 %
USP - Microbiological E Coli (negative) Passes Test
FCC - Assay (Mg(OH)₂) (dried basis) 95.0 - 100.5 %
FCC - Identification Passes Test
FCC - Loss on Drying at 105°C ≤ 2.0 %
FCC - Loss on Ignition 30.0 - 33.0 %
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Calcium Oxide ≤ 1 %
FCC - Alkalies (as SO₄) Passes Test
Appearance (fine, white powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty