Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mineral oil heavy, white stabilized USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Stabilization: Stabilised with vitamin E 10-20 ppm
Management of Change (MOC) category = R

MACR6357-19 MACR6357-19 6357-10 6357-04 MACR6357-10 MACR6357-04
MK635710CS 1176.29 USD InStock
CAMK635719 MK635710 CAMK635710 MK635704 CAMK635704 MK635719
Mineral oil heavy, white stabilized USP, Macron Fine Chemicals™
Mineral oil


Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07263
Order Now
Specification Test Results
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Acidity Passes Test
USP - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
USP - Limit of Sulfur Compounds Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.845 - 0.905
USP - Viscosity at 40 C, mm₂/s 34.5 - 150.0
USP - Readily Carbonizable Substances Passes Test
USP - Solid Paraffin Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty