Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mineral oil light, white NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

MACR6358-10 MACR6358-10 6358-04 MACR6358-19 MACR6358-04 MACR6358-19
MK635810CS 1244.57 USD InStock
CAMK635804 MK635810 CAMK635819 CAMK635810 MK635804 MK635819
Mineral oil light, white NF, Macron Fine Chemicals™
Mineral oil


Boiling Pt: 300…500 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 215 °C (closed cup)
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093
Order Now
Specification Test Results
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Specific Gravity at 25°/25°C 0.818 - 0.880
NF - Viscosity at 40 C, mm₂/s 3.0 - 34.4
NF - Readily Carbonizable Substances Passes Test
NF - Solid Paraffin Passes Test
NF - Acidity Passes Test
NF - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
NF - Limit of Sulfur Compounds Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty