Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silica gel, SilicAR Grade 62, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

MACR6551-02 MACR6551-05 MACR6551-05 MACR6551-02
MK655105EA 1105.23 USD InStock
CAMK655102 MK655105 CAMK655105 MK655102
Silica gel, SilicAR Grade 62, Macron Fine Chemicals™
Silica gel
60 to 200 mesh. For column chromatography.


Formula: SiO₂
MW: 60.08 g/mol
MDL Number: MFCD00011232
CAS Number: 63231-67-4
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Loss on Ignition ≤12 %
On U.S. No. 60 Sieve ≤5 %
On U.S. No. 200 Sieve ≥90 %
Appearance (fine, white powder)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty