Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen carbonate 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Potassium bicarbonate

Management of Change (MOC) category = N

MACR6748-06 MACR6748-20 MACR6748-20
MK674820EA 971.53 USD InStock
MK674820 CAMK674820 MK674806
Potassium hydrogen carbonate 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Potassium hydrogen carbonate


Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 12,07770
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KHCO₃) 99.7 - 100.5 %
ACS - Ammonium (NH₄) ≤0.0005 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤5 ppm
ACS - Magnesium (Mg) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sodium (Na) ≤0.03 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
Appearance (colorless, transparent granules)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty