Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Potassiumchloride

MACR6838-26 MACR6838-20 MACR6838-26
MK683826EA 1891.84 USD InStock
CAMK683820 MK683826 CAMK683826
Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Potassium chloride


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704
Order Now
Specification Test Results
Meets U.S.P Requirements
Meets F.C.C. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (KCl) (by Ag titrn) 99.0 - 100.5 %
USP - Acidity or Alkalinity (no pink color then pink) Passes Test
USP - Bromide (Br)(USP) ≤0.1 %
USP - Calcium and Magnesium Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iodide (I)(USP) ≤0.005 %
USP - Loss on Drying at 105°C ≤1.0 %
USP - Sodium (Na) Passes Test
FCC - Assay (KCl) (by Ag titrn) 99.0 - 100.5 %
FCC - Acidity or Alkalinity (no pink color then pink) Passes Test
FCC - Bromide and/or Iodide (FCC) Passes Test
FCC - Heavy Metals (as Pb)(FCC) ≤5 mg/kg
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying at 105°C ≤1.0 %
FCC - Sodium (Na) Passes Test
Solubility (10 in 50) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty