Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Florisil® 60-100 mesh for pesticide residue analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: TalcFlorisilSilicic acid magnesium saltMagnesium silicate, activated

Management of Change (MOC) category = N

MACR6877-08 MACR6877-08 MACR6877-06 MACR6877-06
MK687706CS 3775.52 USD InStock
MK687706 MK687708 CAMK687708 CAMK687706
Florisil® 60-100 mesh for pesticide residue analysis, Macron Fine Chemicals™
Magnesium silicate
Suitable for use in pesticide residue analysis.


Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 g/cm³
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
GLC Interference Passes Test
On U.S. No. 60 Sieve ≤15 %
Thru U.S. No. 60 Sieve ≥75 %
On U.S. No. 100 Sieve ≥75 %
Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder)
Product may contain black particles that are produced during the manufacturing process.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty