Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen sulfate, fused, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potassium bisulphatePotassium bisulfate

Management of Change (MOC) category = N

MACR7112-04 MACR7112-06 MACR7112-20 MACR7112-20 MACR7112-06
MK7112-20EA 9997.24 USD InStock
CAMK711204 CAMK711206 CAMK7112-20 MK7112-20 MK711206
Potassium hydrogen sulfate, fused, powder, AR® ACS, Macron Fine Chemicals™
Potassium hydrogen sulphate


Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 12,07774
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Acidity (as H₂SO₄) 37.5 - 38.6 %
ACS - Water (H₂O) ≤2.5 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %
ACS - Sodium (Na) ≤0.01 %
ACS - Insoluble Matter ≤0.01 %
ACS - Ammonium Hydroxide Precipitate ≤0.01 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty