Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium sulfate 99.0-100.5% FCC, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = RL

MACR7132-28 MACR7132-28 MACR7132-23
MK7132-28EA 1960.35 USD InStock
CAMK7132-23 CAMK7132-28 MK7132-28
Potassium sulfate 99.0-100.5% FCC, purified, Macron Fine Chemicals™
Potassium sulphate


Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698 °C (1013 hPa)
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 12,07845
Order Now
Specification Test Results
Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Assay (K₂SO₄) 99.0 - 100.5 %
Appearance (white powder) Passes Test
Identification A Passes Test
Identification B Passes Test
Solubility Passes Test
Heavy Metals (as Pb) ≤10 ppm
Lead (Pb) ≤2 mg/kg
Selenium (Se) ≤5 mg/kg
pH 5.5 - 8.5
Reaction Passes Test
Chloride (Cl) ≤0.07 %
Arsenic (As) ≤3 ppm
Calcium (Ca) Passes Test
Magnesium (Mg) ≤0.1 %
Sodium (Na) ≤0.05 %
On U.S. No. 80 Sieve ≤5 %
On U.S. No. 120 Sieve, For Information Only

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty