Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger

Management of Change (MOC) category = N

MACR7180-08 MACR7180-19 MACR7180-04 MACR7180-04 MACR7180-08 MACR7180-19
MK718004CS 3457.3 USD InStock
MK718004 CAMK718008 CAMK718019 MK718019 MK718008 CAMK718004
Pyridine ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Pyridine


Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 12,08153
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₅H₅N) (by GC, corrected for water) ≥99.0 %
ACS - Solubility in H₂O Passes Test
ACS - Residue after Evaporation ≤0.002 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Ammonia (as NH₃) ≤0.002 %
ACS - Copper (Cu) ≤5 ppm
ACS - Reducing Substances Passes Test
Appearance (clear, colorless liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty