Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate 99.0-100.5% (by acidimetry on anhydrous substance), powder USP, FCC for haemodialysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

MACR7396-04 MACR7396-88 MACR7396-08 MACR7396-06 MACR7396-88 MACR7396-26 MACR7396-20 MACR7396-06 MACR7396-08 MACR7396-26 MACR7396-20 MACR7396-08
MK739620EA 581.65 USD InStock
CAMK739606 CAJT7396-08 CAMK739626 MK739620 MK739688 JT7396-08 MK7396-08 CAMK739620 MK739604 MK739626 MK739606 CAMK739688
Sodium hydrogen carbonate 99.0-100.5% (by acidimetry on anhydrous substance), powder USP, FCC for haemodialysis, Macron Fine Chemicals™
Sodium hydrogen carbonate
pH adjusters are used to adjust the pH of formulations.


Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 12,08726
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now
Specification Test Results
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
USP - Aluminum (Al) ≤2 ppm
USP - Arsenic (As) ≤2 ppm
USP - Calcium (Ca) ≤0.01 %
USP - Carbonate (CO₃) ≤0.23 %
USP - Chloride (Cl) ≤0.015 %
USP - Copper (Cu) ≤1 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances Passes Test
USP - Iron (Fe) ≤5 ppm
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Magnesium (Mg) ≤0.004 %
USP - Normal Carbonate Passes Test
USP - Limit of Organics ≤0.01 %
FCC - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
FCC - Ammonia (as NH₃) Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty