Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium bicarbonate

Management of Change (MOC) category = N

MACR7412-20 MACR7412-06 MACR7412-09 7412-12 MACR7412-20 MACR7412-12 MACR7412-28 MACR7412-28 MACR7412-09
MK741212CS 304.19 USD InStock
MK741212 CAMK7412-28 MK741220 CAMK7412-09 JT7412-09 CAMK741220 MK7412-28 MK741206 CAMK741212
Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Sodium hydrogen carbonate


Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 12,08726
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NaHCO₃) (by acidimetry) 99.7 - 100.3 %
ACS - Ammonium (NH₄) ≤5 ppm
ACS - Calcium (Ca) ≤0.02 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.015 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty