Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen sulfate monohydrate, white crystals, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium bisulfate monohydrateSodium bisulphate monohydrate

Management of Change (MOC) category = N

MACR7432-20 MACR7432-12 MACR7432-20 MACR7432-12
MK743220EA 5083.04 USD InStock
CAMK743220 MK743220 MK743212 CAMK743212
Sodium hydrogen sulfate monohydrate, white crystals, AR®, Macron Fine Chemicals™
Sodium hydrogen sulphate monohydrate


Formula: NaHSO₄·H₂O
MW: 138.08 g/mol
Melting Pt: 183 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 13,08658
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Solubility (10 in 50) Passes Test
Chloride (Cl) ≤0.001 %
Acidity (as H₂SO₄) 35 - 37 %
Arsenic (As) ≤0.0001 %
Insoluble Matter and NH₄OH Precipitate ≤0.005 %
Calcium and Magnesium Precipitate ≤0.005 %
Heavy Metals (as Pb) ≤0.0005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Appearance (white crystals)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty