Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 95.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = R

MACR7680-88 MACR7680-04 MACR7680-28 MACR7680-20 MACR7680-06 MACR7680-88 MACR7680-28 MACR7680-25 MACR7680-04 MACR7680-25 MACR7680-20 7680-26 MACR7680-06
MK768028EA 1289.02 USD InStock
MK768028 MK768006 MK768004 CAMK768020 CAMK768088 MK768020 MK7680-26 CAMK7680-25 CAMK768028 CAMK768006 MK768088 CAJT7680-4 MK7680-25
Sodium hydroxide 95.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
GMP Manufactured Product
Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Appearance (white hygroscopic pellets) Passes Test
NF - Insoluble Substances and Organic Matter Passes Test
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Potassium (K) ≤0.5 %
NF - Sodium Carbonate (Na₂CO₃) ≤3.0 %
FCC - Assay (Total alkali as NaOH) (by acidimetry) 95.0 - 100.5 %
FCC - Identification Passes Test
FCC - Insoluble Substances and Organic Matter Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤0.1 mg/kg
FCC - Arsenic (As) ≤3 mg/kg
FCC - Sodium Carbonate (Na₂CO₃) ≤3.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty