Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 99.0-100.5% (dried basis), granular, GenAR® USP for haemodialysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: HaliteSea saltNaClSaline

Management of Change (MOC) category = R

MACR7713-B4 MACR7713-60 MACR7713-B3 MACR7713-45 MACR7713-75 MACR7713-25 MACR7713-F9 MACR7713-73 MACR7713-73 MACR7713-88 MACR7713-60 MACR7713-75 MACR7713-B4 MACR7713-88 MACR7713-70 MACR7713-70 MACR7713-30 MACR7713-06 MACR7713-04 MACR7713-04 MACR7713-45 MACR7713-B3 MACR7713-20 MACR7713-20 MACR7713-25 MACR7713-B4 MACR7713-06 MACR7713-30 MACR7713-29 MACR7713-F9 MACR7713-29 7713-70
MK771306CS 701.29 USD InStock
CAMK7713-60 MK771306 CAMK771388 MK771329 MK7713-75 MK771304 CAJT7713-45 CAMK771320 MK771325 MK771388 CAMK7713-F9 MK7713-70 JT7713-B3 JT7713-B4 CAJT7713-73 CAMK7713-70 JT7713-45 CAMK7713-75 CAJT7713-30 MK7713-F9 MK7713-60 CAJT7713-B3 TXL7713-B4BYE CAJT7713-B4 JT7713-30 MK771320 CAMK771329 JT7713-73 CAMK771306 JT7713-70 CAMK771304 CAMK771325
Sodium chloride 99.0-100.5% (dried basis), granular, GenAR® USP for haemodialysis, Macron Fine Chemicals™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
Meets F.C.C. Requirements
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (NaCl)(USP) (dried basis) 99.0 - 100.5 %
USP - Appearance of Solution Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Bromide (Br) ≤100 ppm
USP - Loss on Drying at 105°C ≤0.5 %
USP - Iodide (I) Passes Test
USP - Aluminum (Al) ≤0.2 ppm
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
USP - Arsenic (As) ≤1 ppm
USP - Barium (Ba) Passes Test
USP - Ferrocyanide Passes Test
USP - Sulfate (SO₄) ≤200 ppm
USP - Iron (Fe) ≤2 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤0.01
USP - Phosphate (PO₄) ≤25 ppm
USP - Potassium (K) ≤500 ppm
EP/BP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Bromide (Br) ≤100 ppm
EP/BP - Ferrocyanide Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Iodide (I) Passes Test
EP/BP - Nitrite (NO₂) ≤0.01
EP/BP - Phosphate (PO₄) ≤25 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Aluminum (Al) ≤0.2 ppm
EP/BP - Arsenic (As) ≤1 ppm
EP/BP - Barium (Ba) Passes Test
EP/BP - Iron (Fe) ≤2 ppm
EP/BP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
EP/BP - Potassium (K) ≤500 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Endotoxin Concentration, <5 IU/g Passes Test
Endotoxin Concentration (EU/g) ≤2.5
JP - Assay 99.0 - 100.5 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Bromide (Br) Passes Test
JP - Iodide (I) Passes Test
JP - Ferrocyanide Passes Test
JP - Iron (Fe) Passes Test
JP - Heavy Metals (as Pb) ≤3 ppm
JP - Barium (Ba) Passes Test
JP - Magnesium and Alkaline Earth Metals Passes Test
JP - Phosphate (PO₄) Passes Test
JP - Sulfates (as SO₄) Passes Test
JP - Arsenic (As) ≤2 ppm
JP - Loss on Drying at 105°C ≤0.5 %
FCC - Assay (as NaCl) (FCC) (ignited basis) 99.0 - 100.5 %
FCC - Arsenic (As) ≤1 ppm
FCC - Calcium and Magnesium (FCC) ≤0.35 %
FCC - Heavy Metals (as Pb)(FCC) ≤2 mg/kg
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying at 105°C ≤0.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty