Synonyms:
Guanidinium chloride
, Guanidine hydrochloride
, Guanidine chloride
, Guanadine hydrochloride
Management of Change (MOC) category = N
MACR7716-04
MACR7716-87
MACR7716-04
MACR7716-87
MACR7716-88
MACR7716-88
MK771604CS
2306.25
USD
InStock
MK771604
MK771687
MK771688
CAMK771604
CAMK771688
CAMK771687
Guanidinium hydrochloride ≥99.5%, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride
Tested for absence of detectable aldehydes and ammeline. DNase-, RNase-, and protease-free.