Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 95.0-100.5%, pellets, GenAR® NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = R

MACR7772-06 MACR7772-88 MACR7772-26 MACR7772-30 MACR7772-30 MACR7772-28 MACR7772-25 MACR7772-25 MACR7772-30 MACR7772-26 MACR7772-06 MACR7772-28 MACR7772-88
MK7772-30CS 1977.74 USD InStock
MK7772-30 CAMK7772-30 MK777288 MK777226 MK777225 MK777206 CAMK777288 MK777228 CAMK777226 CAMK777225 CAMK777206 CAMK777228 JT7772-30
Sodium hydroxide 95.0-100.5%, pellets, GenAR® NF, Macron Fine Chemicals™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Meets F.C.C. Requirements
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Food GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Insoluble Substances and Organic Matter Passes Test
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Potassium (K) ≤0.5 %
NF/FCC - Carbonate (as Na₂CO₃) ≤3.0 %
FCC - Assay (Total alkali as NaOH) 95.0 - 100.5 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg)(mg/kg) ≤0.1
FCC - Arsenic (As)(mg/kg) ≤3
FCC - Identification Passes Test
FCC - Insoluble Substances and Organic Matter Passes Test
BP/EP - Assay (Total alkali as NaOH) 97.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤2.0 %
JP - Mercury (Hg) Passes Test
Endotoxin Concentration (EU/g) ≤5
Appearance (white hygroscopic pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty