Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium peroxide ≥93.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: di-Sodium peroxide

Management of Change (MOC) category = N

MACR7864-04 MACR7864-04
MK786404CS 4096.06 USD InStock
CAMK786404 MK786404
Sodium peroxide ≥93.0%, granular, AR® ACS, Macron Fine Chemicals™
Sodium peroxide
Granular.


Formula: Na₂O₂
MW: 77.98 g/mol
Melting Pt: 660 °C
Density: 2.805 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 12,08800
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂O₂) (by KMnO₄ titrn) ≥93.0 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sulfate (SO₄) ≤0.001 %
Appearance (white to pale yellow, hygroscopic granules)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty