Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium succinate hexahydrate, granular, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium succinate dibasic hexahydrateSuccinic acid disodium salt hexahydrateSodium succinate hexahydrateSuccinic acid disodium hexahydrateSodium Succinate, HexahydrateDisodium succinate hexahydrate1,4-Butanedioic acid disodium salt hexahydrate

Management of Change (MOC) category = N

MACR7980-05 MACR7980-03 MACR7980-05 MACR7980-03
MK798005CS 4374.24 USD InStock
CAMK798005 CAMK798003 MK798005 MK798003
di-Sodium succinate hexahydrate, granular, Macron Fine Chemicals™
di-Sodium succinate hexahydrate


Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 300 °C
Density: 1.529 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay, For Information Only
Chloride (Cl) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
pH of 0.4M Solution 6.0 - 9.0
Solubility Passes Test
Substances Reducing Permanganate Passes Test
Sulfate (SO₄) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty