Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate, anhydrous ≥99.0% (dry basis) USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Sulphuric acid disodium saltBisodium sulphateSulfuric acid disodium saltBisodium sulfateDisodium sulphateDisodium sulfate

Management of Change (MOC) category = R

MACR8028-QM MACR8028-20 MACR8028-F9 MACR8028-12 8028-88 MACR8028-88 MACR8028-12 MACR8028-06 MACR8028-W3 MACR8028-F9 MACR8028-B1 MACR8028-B1 MACR8028-65 MACR8028-20 MACR8028-06 MACR8028-QM MACR8028-65 MACR8028-W3
MK8028QMEA 88.56 USD InStock
MK8028QM MK8028W3 MK802865 MK802820 CAMK8028QM MK802812 CAMK8028W3 MK802888 CAMK802888 CAMK802812 CAMK802820 CAMK802865 CAMK8028-B1 MK8028-B1 CAMK802806 MK8028F9 MK802806 CAMK8028F9
Sodium sulfate, anhydrous ≥99.0% (dry basis) USP, Macron Fine Chemicals™
Sodium sulphate
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.


Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (Na₂SO₄) (dried basis) ≥ 99.0 %
USP - Acidity or Alkalinity (mL) ≤ 0.50
USP - Chloride (Cl) ≤ 0.005 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤ 0.01 %
USP - Loss on Drying at 105°C ≤ 0.5 %
Thru U.S. No. 50 Sieve, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.
Appearance - Colorless to slightly colored crystalline solid.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty