Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate pentahydrate 99.5-101.0%, crystals, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

MACR8100-04 MACR8100-20 MACR8100-06 MACR8100-04 MACR8100-20 MACR8100-06
MK810020EA 501.49 USD InStock
MK810020 CAMK810006 CAMK810004 MK8100-04 MK810006 CAMK810020
Sodium thiosulfate pentahydrate 99.5-101.0%, crystals, AR®, Macron Fine Chemicals™
Sodium thiosulphate pentahydrate


Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (Na₂S₂O₃·5H₂O) 99.5 - 101.0 %
Appearance (colorless, transparent crystals) Passes Test
Insoluble Matter ≤0.005 %
Nitrogen Compounds (as N) ≤0.002 %
pH of 5% Solution at 25°C 6.0 - 8.4
Solubility Passes Test
Sulfate and Sulfite (as SO₄) ≤0.1 %
Sulfide (S) Passes Test
Water (H₂O)(by Karl Fischer titrn), For Information Only

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty