Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Reactive blue 4, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

MACR8132-03
MK813203EA 1300.08 USD InStock
MK813203
Reactive blue 4, OR, Macron Fine Chemicals™
Reactive blue 4


Formula: C₂₃H₁₄Cl₂N₆O₈S₂
MW: 637.44 g/mol
MDL Number: MFCD00001218
CAS Number: 13324-20-4
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Identification Passes Test
UV Absorbance at 280 nm and 253 nm Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty