Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) chloride dihydrate 98.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Stannous chloride dihydrateStannous Chloride Dihydrate

MACR8176-04 MACR8176-04 MACR8176-06 MACR8176-20 MACR8176-06 MACR8176-20
MK8176-04CS 2032.41 USD InStock
CAMK817620 MK8176-04 CAMK8176-04 MK817606 CAMK817606 MK817620
Tin(II) chloride dihydrate 98.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™
Tin (II) chloride dihydrate


Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 12,08939
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
ACS - Assay (SnCl₂·2H₂O) 98.0 - 103.0 %
ACS - Mercury (Hg) ≤0.050 ppm
ACS - Solubility in HCl Passes Test
ACS - Sodium (Na) ≤0.01 %
ACS - Potassium (K) ≤0.005 %
ACS - Iron (Fe) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.01 %
Appearance: colorless to slightly yellow (but definitely not green) crystals

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty